Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP13A2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$192.47 - $517.59
Specifications
| Antigen | ATP13A2 |
|---|---|
| Concentration | 0.75 mg/mL |
| Dilution | Western Blot 2ug/ ml, Simple Western 1:100, Immunohistochemistry 10 ug/ml |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11041486T
![]() |
Novus Biologicals
NB11041486SS |
0.025 mL |
Each for $192.47
|
|
|||||
NB11041486
![]() |
Novus Biologicals
NB11041486 |
0.1 mL |
Each for $517.59
|
|
|||||
Description
ATP13A2 Polyclonal specifically detects ATP13A2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
| ATP13A2 | |
| Western Blot 2ug/ ml, Simple Western 1:100, Immunohistochemistry 10 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Cancer, Neurodegeneration, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| ATPase type 13A2, EC 3.6.3, EC 3.6.3.-, EC 3.6.3.5, EC 3.6.3.8, FLJ26510, HSA9947, KRPPD, PARK9putative ATPase, Parkinson disease (autosomal recessive) 9 (Kufor-Rakeb syndrome), probable cation-transporting ATPase 13A2 | |
| ATP13A2 | |
| IgG | |
| Affinity Purified |
| 0.75 mg/mL | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q9NQ11 | |
| 23400 | |
| A synthetic peptide made to an internal region of the human ATP13A2 protein (within residues 200-300). [Swiss-Prot# Q9NQ11] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title