Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP1B4 Antibody (108), FITC, Novus Biologicals™
SDP

Catalog No. NB070444 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB070444 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB070444 Supplier Novus Biologicals Supplier No. NBP290158F
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ATP1B4 Monoclonal specifically detects ATP1B4 in Human samples. It is validated for ELISA.

Specifications

Antigen ATP1B4
Applications ELISA
Classification Monoclonal
Clone 108
Conjugate FITC
Dilution ELISA
Formulation PBS
Gene Alias ATPase, (Na+)/K+ transporting, beta 4 polypeptide, ATPase, Na+/K+ transporting, beta 4 polypeptide, K-ATPase beta-m subunit, K-ATPase subunit beta-m, protein ATP1B4, X, X/potassium-transporting ATPase subunit beta-m
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ATP1B4 (mature form, NP_036201.1; Tyr 132-Thr 357).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23439
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.