Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP5G2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ATP5G2 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2539
![]() ![]() |
Proteintech Group Inc
197851AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is specific to ATP5G2.
ATP5G2, also named as ATPase protein 9 and ATPase subunit c, belongs to the ATPase C chain family. Mitochondrial membrane ATP synthase (F1F0 ATP synthase or Complex V) produces ATP from ADP in the presence of a proton gradient across the membrane which is generated by electron transport complexes of the respiratory chain. ATP5G2 is the major protein stored in the storage bodies of animals or humans affected with ceroid lipofuscinosis (Batten disease).Specifications
ATP5G2 | |
Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P56383, Q06055 | |
517, 67942 | |
Peptide | |
Primary | |
-20°C | |
ATP5G2 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ATP synthase proteolipid P2, ATP5G2, ATPase protein 9, ATPase subunit c | |
ATP5G2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title