Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP5H Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB044388 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB044388 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB044388 Supplier Novus Biologicals Supplier No. NBP297512JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATP5H Polyclonal specifically detects ATP5H in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ATP5H
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ATP synthase D chain, mitochondrial, ATP synthase subunit d, mitochondrial, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit d, ATP synthase, H+ transporting, mitochondrial F1F0, subunit d, ATP synthase, H+ transporting, mitochondrial Fo complex, subunit d, ATP5JD, ATPase subunit d, ATPQ, My032 protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ATP5H fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10476
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.