Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP5J2 Antibody, Novus Biologicals™
SDP

Catalog No. NB403552 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB403552 25 μL
NBP257076 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB403552 Supplier Novus Biologicals Supplier No. NBP25707625UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATP5J2 Polyclonal specifically detects ATP5J2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ATP5J2
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 μg/mL
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias ATP synthase f chain, mitochondrial, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit f, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit f, isoform 2, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F2, ATP synthase, H+ transporting, mitochondrial Fo complex, subunit F2, ATP5JLATP synthase subunit f, mitochondrial, F1F0-type ATPase subunit f, F1Fo-ATP synthase complex Fo membrane domain f subunit, F1Fo-ATPase, F1Fo-ATPase synthase f subunit
Gene Symbols ATP5MF
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FGAFQRGYYRYYNKYINVKK
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9551
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.