Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP5O Antibody, Novus Biologicals™
SDP

Catalog No. NBP131646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP131646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP131646 Supplier Novus Biologicals Supplier No. NBP131646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATP5O Polyclonal specifically detects ATP5O in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ATP5O
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal
Gene Alias ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit, human ATP synthase OSCP subunit, oligomycin sensitivity conferring protein10ATPOATP synthase subunit O, mitochondrial, Oligomycin sensitivity conferral protein, oligomycin sensitivity conferring protein, OSCPhuman ATP synthase OSCP subunit
Gene Symbols ATP5PO
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ATP5O. The exact sequence is proprietary.
Molecular Weight of Antigen 23 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 539
Test Specificity Predicted cross-reactivity based on sequence identity: Monkey (98%), Rabbit (88%), and Bovine (83%). Not yet tested in any other species.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.