Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP6V0A1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$416.50 - $682.00
Specifications
Antigen | ATP6V0A1 |
---|---|
Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ATP6V0A1 Polyclonal specifically detects ATP6V0A1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
ATP6V0A1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
535 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:VQFRDLNPDVNVFQRKFVNEVRRCEEMDRKLRFVEKEIRKANIPIMDTGENPEVPFPRDMIDLEANFEKIENELKEINTNQEALKRNFLELTELK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human, Mouse | |
a1, ATP6N1, ATP6N1AATPase, H+ transporting, lysosomal non-catalytic accessory protein 1(110/116kD), ATP6V0, ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalyticaccessory protein 1A (110/116kD), ATPase, H+ transporting, lysosomal V0 subunit a isoform 1, ATPase, H+ transporting, lysosomal V0 subunit a1, Clathrin-coated vesicle/synaptic vesicle proton pump 116 kDa subunit, DKFZp781J1951, Stv1, Vacuolar adenosine triphosphatase subunit Ac116, Vacuolar proton pump subunit 1, vacuolar proton pump, subunit 1, vacuolar proton translocating ATPase 116 kDa subunit A, Vacuolar proton translocating ATPase 116 kDa subunit a isoform 1, vacuolar-type H(+)-ATPase 115 kDa subunit, V-ATPase 116 kDa, V-ATPase 116 kDa isoform a1, Vph1, VPP1H(+)-transporting two-sector ATPase, 116 kDa accessory protein A1, V-type proton ATPase 116 kDa subunit a, V-type proton ATPase 116 kDa subunit a isoform 1 | |
ATP6V0A1 | |
IgG | |
Affinity Purified | |
Specificity of human ATP6V0A1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title