Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V0C Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB133540 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB133540 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB133540 Supplier Novus Biologicals Supplier No. NBP306206C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATP6V0C Polyclonal specifically detects ATP6V0C in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ATP6V0C
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ATP6CATPase, H+ transporting, lysosomal (vacuolar proton pump) 16kD, ATP6LVPPC, ATPase, H+ transporting, lysosomal 16kDa, V0 subunit c, ATPL, H(+)-transporting two-sector ATPase, 16 kDa subunit, vacuolar ATP synthase 16 kDa proteolipid subunit, vacuolar H+ ATPase proton channel subunit, Vacuolar proton pump 16 kDa proteolipid subunit, VATL, V-ATPase 16 kDa proteolipid subunit, Vma3, V-type proton ATPase 16 kDa proteolipid subunit
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ATP6V0C.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 527
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.