Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V0D1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB324734 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB324734 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB324734 Supplier Novus Biologicals Supplier No. NBP299561MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATP6V0D1 Polyclonal antibody specifically detects ATP6V0D1 in Human samples. It is validated for Western Blot

Specifications

Antigen ATP6V0D1
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias 32 kDa accessory protein, ATP6D, ATP6DV, ATP6V0, ATPase, H+ transporting, lysosomal (vacuolar proton pump), member D, ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d isoform 1, ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d1, FLJ43534, H(+)-transporting two-sector ATPase, subunit D, P39, Vacuolar proton pump subunit d 1, V-ATPase 40 kDa accessory protein, V-ATPase AC39 subunit, V-ATPase, subunit D, VATX, Vma6, VPATPDV-ATPase subunit d 1, V-type proton ATPase subunit d 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ATP6V0D1.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9114
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.