Learn More
Description
Specifications
Specifications
| Antigen | ATP6V0E2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.2-1 ug/ml |
| Formulation | PBS and 2% Sucrose with 0.09% Sodium Azide |
| Gene Alias | ATPase, H+ transporting V0 subunit e2, chromosome 7 open reading frame 32, H+ transporting V0 subunit E isoform 2-like (rat), Lysosomal 9 kDa H(+)-transporting ATPase V0 subunit e2 |
| Gene Symbols | ATP6V0E2 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptides corresponding to ATP6V0E2(ATPase, H+ transporting V0 subunit e2) The peptide sequence was selected from the middle region of ATP6V0E2. Peptide sequence TVAPLSLTTPSSGPSPTQLCLVTSSLLLAPRDPDPQGLPGSWKSSQSSQP. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
