Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V1B1 Antibody (OTI1H6), FITC, Novus Biologicals™
SDP

Catalog No. NBP270237F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270237F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270237F Supplier Novus Biologicals Supplier No. NBP270237F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATP6V1B1 Monoclonal specifically detects ATP6V1B1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ATP6V1B1
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1H6
Conjugate FITC
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias ATP6B158kD subunit, ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1, Endomembrane proton pump 58 kDa subunit, H+-ATPase beta 1 subunit, vacuolar proton pump 3, Vacuolar proton pump subunit B 1, vacuolar proton pump, subunit 3, VATBMGC32642, V-ATPase B1 subunit, V-ATPase subunit B 1, Vma2, V-type proton ATPase subunit B, kidney isoform
Gene Symbols ATP6V1B1
Host Species Mouse
Immunogen Full length recombinant protein of human ATP6V1B1 (NP_001683) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 525.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.