Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V1B2 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB101286 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB101286 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB101286 Supplier Novus Biologicals Supplier No. NBP297188UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATP6V1B2 Polyclonal specifically detects ATP6V1B2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ATP6V1B2
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias 000 subunit, 56/58kD, isoform 2, ATP6B2ATPase, H+ transporting, lysosomal (vacuolar proton pump), beta polypeptide, ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B2, Endomembrane proton pump 58 kDa subunit, H+ transporting two-sector ATPase, HO57ATP6B1B2, vacuolar H+-ATPase 56, Vacuolar proton pump subunit B 2, VATB, V-ATPase B2 subunit, V-ATPase subunit B 2, Vma2, VPP3ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B, isoform 2, V-type proton ATPase subunit B, brain isoform
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ATP6V1B2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 526
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.