Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V1C1 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB325336 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325336 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325336 Supplier Novus Biologicals Supplier No. NBP299458F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATP6V1C1 Polyclonal antibody specifically detects ATP6V1C1 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen ATP6V1C1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias ATP6CH+-transporting ATPase chain C, vacuolar, ATP6Dsubunit C of vacuolar proton-ATPase V1 domain, ATPase, H+ transporting, lysosomal (vacuolar proton pump) 42kD, ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C, isoform 1, ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C1, FLJ20057, H(+)-transporting two-sector ATPase, subunit C, vacuolar ATP synthase subunit C, vacuolar proton pump C subunit, Vacuolar proton pump subunit C 1, vacuolar proton pump, 42-kD subunit, vacuolar proton-ATPase, subunit C, VI domain, VATCH+ -ATPase C subunit, V-ATPase C subunit, V-ATPase subunit C 1, Vma5, V-type proton ATPase subunit C 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ATP6V1C1.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 528
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.