Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP6V1C2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | ATP6V1C2 |
---|---|
Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ATP6V1C2 Polyclonal antibody specifically detects ATP6V1C2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ATP6V1C2 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
ATP6C2, ATPase, H+ transporting, lysosomal 42kD, V1 subunit C isoform 2, ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C isoform 2, ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C2, Vacuolar proton pump subunit C 2, V-ATPase C2 subunit, V-ATPase subunit C 2, VMA5, V-type proton ATPase subunit C 2 | |
This antibody was developed against a recombinant protein corresponding to amino acids: YPVKQPLVSVVDTIAKQLAQIEMDLKSRTAAYNTLKTNLENLEKKSMGNLFTRTLSDIVSKED | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.2) and 40% Glycerol | |
245973 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title