Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V1D Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB354527 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB354527 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB354527 Supplier Novus Biologicals Supplier No. NBP297189PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATP6V1D Polyclonal antibody specifically detects ATP6V1D in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen ATP6V1D
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ATP6M, ATPase, H+ transporting lysosomal, member M, ATPase, H+ transporting, lysosomal (vacuolar proton pump), ATPase, H+ transporting, lysosomal 34kDa, V1 subunit D, H(+)-transporting two-sector ATPase, subunit M, vacuolar ATP synthase subunit D, vacuolar H-ATPase subunit D, vacuolar proton pump D subunit, vacuolar proton pump delta polypeptide, Vacuolar proton pump subunit D, vacuolar proton-ATPase subunit D, VATDV-ATPase D subunit, V-ATPase 28 kDa accessory protein, V-ATPase subunit D, VMA8, V-type proton ATPase subunit D
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ATP6V1D fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51382
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.