Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP6V1E2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | ATP6V1E2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1546020
![]() |
Novus Biologicals
NBP15460120UL |
20 μL |
Each for $206.00
|
|
|||||
NBP154601
![]() |
Novus Biologicals
NBP154601 |
100 μL |
Each for $487.50
|
|
|||||
Description
ATP6V1E2 Polyclonal specifically detects ATP6V1E2 in Human samples. It is validated for Western Blot.Specifications
ATP6V1E2 | |
Polyclonal | |
Rabbit | |
Q96A05 | |
90423 | |
Synthetic peptides corresponding to ATP6V1E2(ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E2) The peptide sequence was selected from the middle region of ATP6V1E2. Peptide sequence LMSTMRNQARLKVLRARNDLISDLLSEAKLRLSRIVEDPEVYQGLLDKLV | |
Primary | |
26 kDa |
Western Blot | |
Unconjugated | |
RUO | |
ATP6E1V-ATPase subunit E 2, ATP6EL2, ATP6V1EL2, ATPase, H+ transporting, lysosomal (vacuolar proton pump) 31kD-like 2, ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E isoform 2, ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E2, MGC9341, Vacuolar proton pump subunit E 2, vacuolar-type proton-translocating ATPase subunit E1, VMA4, V-type proton ATPase subunit E 2 | |
ATP6V1E2 | |
IgG | |
This product is specific to Subunit or Isoform: E 2. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title