Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V1E2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP215521 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215521 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215521 Supplier Novus Biologicals Supplier No. NBP215521
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATP6V1E2 Polyclonal specifically detects ATP6V1E2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen ATP6V1E2
Applications Western Blot
Classification Polyclonal
Concentration 0.69mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:20000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q96A05
Gene Alias ATP6E1V-ATPase subunit E 2, ATP6EL2, ATP6V1EL2, ATPase, H+ transporting, lysosomal (vacuolar proton pump) 31kD-like 2, ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E isoform 2, ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E2, MGC9341, Vacuolar proton pump subunit E 2, vacuolar-type proton-translocating ATPase subunit E1, VMA4, V-type proton ATPase subunit E 2
Gene Symbols ATP6V1E2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ATP6V1E2. The exact sequence is proprietary.
Molecular Weight of Antigen 26 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90423
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.