Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V1H Antibody (HL1316) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB295245 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295245 Supplier Novus Biologicals Supplier No. NBP325372
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ATP6V1H Monoclonal antibody specifically detects ATP6V1H in Human, Mouse, Drosophila samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen ATP6V1H
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HL1316
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ATPase, H+ transporting, lysosomal 50/57kD, V1 subunit H, ATPase, H+ transporting, lysosomal 50/57kDa, V1 subunit H, CGI-11, MSTP042, NBP1, Nef-binding protein 1, Protein VMA13 homolog, SFD, SFDalpha, SFDbeta, vacuolar ATP synthase subunit H, vacuolar ATPase subunit H, vacuolar proton pump H subunit, Vacuolar proton pump subunit H, Vacuolar proton pump subunit SFD, V-ATPase 50/57 kDa subunits, V-ATPase H subunit, V-ATPase subunit H, VMA13, V-type proton ATPase subunit H
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ATP6V1H. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51606
Target Species Human, Mouse, Drosophila
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.