Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP7A Antibody (S60-4), Novus Biologicals™
SDP

Catalog No. NBP259376 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259376 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259376 Supplier Novus Biologicals Supplier No. NBP259376
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

ATP7A Monoclonal specifically detects ATP7A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen ATP7A
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S60-4
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin
Formulation PBS pH7.4, 50% glycerol with 0.09% Sodium Azide
Gene Alias ATPase, Cu++ transporting, alpha polypeptide, Copper pump 1, copper-transporting ATPase 1, Cu++-transporting P-type ATPase, DSMAX, EC 3.6.3, EC 3.6.3.4, MC1, Menkes disease-associated protein, Menkes syndrome, MK, MNKFLJ17790, OHS, SMAX3
Gene Symbols ATP7A
Host Species Mouse
Immunogen Synthetic peptide amino acids 42-61 (cytoplasmic C-terminus) of human Copper- transporting ATPase1
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 538
Test Specificity Detects the extracellular domain of LRP4 215kDa. Degradation product can be seen at approx 150kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.