Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP7b Antibody (36D12), Novus Biologicals™
SDP

Catalog No. NB298000 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298000 50 μL
NB298001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298000 Supplier Novus Biologicals Supplier No. NBP32648850UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ATP7b Monoclonal antibody specifically detects ATP7b in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen ATP7b
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 36D12
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias ATPase, Cu(2+)- transporting, beta polypeptide, ATPase, Cu++ transporting, beta polypeptide, Copper pump 2, copper-transporting ATPase 2, EC 3.6.3, EC 3.6.3.4, PWD, WC1, WD, Wilson disease-associated protein, WNDATPase, Cu++ transporting, beta polypeptide (Wilson disease)
Host Species Rabbit
Immunogen A synthesized peptide derived from Human ATP7b [UniProt P35670]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 540
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.