Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP7b Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB100361R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100361R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100361R Supplier Novus Biologicals Supplier No. NB100361R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATP7b Polyclonal specifically detects ATP7b in Human, Mouse (Negative), Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen ATP7b
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P35670
Gene Alias ATPase, Cu(2+)- transporting, beta polypeptide, ATPase, Cu++ transporting, beta polypeptide, Copper pump 2, copper-transporting ATPase 2, EC 3.6.3, EC 3.6.3.4, PWD, WC1, WD, Wilson disease-associated protein, WNDATPase, Cu++ transporting, beta polypeptide (Wilson disease)
Gene Symbols ATP7B
Host Species Rabbit
Immunogen A synthetic peptide made to an internal sequence near the C-terminus of human ATP7b. [UniProt# P35670]
Molecular Weight of Antigen 165 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 540
Test Specificity Human and rat. Does not appear to work in mouse.
Target Species Human, Mouse (Negative), Mouse (Negative), Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.