Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP7b Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 3 publications

Supplier:  Novus Biologicals NB100361SS

 View more versions of this product

Catalog No. NB100361SS


Only null left
Add to Cart

Description

Description

ATP7b Polyclonal specifically detects ATP7b in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

ATP7b
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
ATPase, Cu(2+)- transporting, beta polypeptide, ATPase, Cu++ transporting, beta polypeptide, Copper pump 2, copper-transporting ATPase 2, EC 3.6.3, EC 3.6.3.4, PWD, WC1, WD, Wilson disease-associated protein, WNDATPase, Cu++ transporting, beta polypeptide (Wilson disease)
Rabbit
165 kDa
0.025 mL
Cancer, Signal Transduction
540
Human, Rat, Mouse (Negative)
IgG
Western Blot, Immunocytochemistry, Immunofluorescence
1 mg/mL
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
P35670
ATP7B
A synthetic peptide made to an internal sequence near the C-terminus of human ATP7b. [UniProt# P35670]
Affinity Purified
RUO
Primary
Human and rat. Does not appear to work in mouse.
Store at -20°C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only