Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP7b Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$206.50 - $499.50
Specifications
Antigen | ATP7b |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100361SS
![]() |
Novus Biologicals
NB100361SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB100361
![]() |
Novus Biologicals
NB100361 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
ATP7b Polyclonal specifically detects ATP7b in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ATP7b | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Polyclonal | |
Rabbit | |
Cancer, Signal Transduction | |
PBS with 0.02% Sodium Azide | |
ATPase, Cu(2+)- transporting, beta polypeptide, ATPase, Cu++ transporting, beta polypeptide, Copper pump 2, copper-transporting ATPase 2, EC 3.6.3, EC 3.6.3.4, PWD, WC1, WD, Wilson disease-associated protein, WNDATPase, Cu++ transporting, beta polypeptide (Wilson disease) | |
ATP7B | |
IgG | |
Affinity Purified | |
Human and rat. Does not appear to work in mouse. |
1 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Rat, Mouse (Negative) | |
P35670 | |
540 | |
A synthetic peptide made to an internal sequence near the C-terminus of human ATP7b. [UniProt# P35670] | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. | |
165 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title