Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP8B3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB166197 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166197 100 μg
NB166198 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166197 Supplier Novus Biologicals Supplier No. NBP315551100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP8B3 Polyclonal antibody specifically detects ATP8B3 in Human samples. It is validated for Western Blot

Specifications

Antigen ATP8B3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias probable phospholipid-transporting ATPase IK, ATPase, aminophospholipid transporter, class I, type 8B, member 3, ATPIK
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 600-900 of human ATP8B3 (NP_620168.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 148229
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.