Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP8B4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | ATP8B4 |
---|---|
Concentration | 0.2mg/mL |
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Description
ATP8B4 Polyclonal specifically detects ATP8B4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ATP8B4 | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
ATPase class I type 8B member 4, ATPase, class I, type 8B, member 4, ATPIM, EC 3.6.3, EC 3.6.3.1, FLJ25418, KIAA1939, potential phospholipid-transporting ATPase IM, probable phospholipid-transporting ATPase IM | |
ATP8B4 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
0.2mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
79895 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:CSINGRIYGEVHDDLDQKTEITQEKEPVDFSVKSQADREFQFFDHNLMESIKMGDPKVHEFLR | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title