Learn More
Specifications
Specifications
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Gene ID (Entrez) | 1769 |
| Gene Symbol | DNAH8 |
| Quantity | 100 Assays |
| Synonym | ATPase, axonemal, dynein, axonemal, heavy chain 8, dynein, axonemal, heavy polypeptide 8, FLJ25850, FLJ36115, FLJ36334, hdhc9 |
| Research Category | Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers |
| Species | Non-species specific |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
