Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATPase Antibody, Novus Biologicals™
SDP

Catalog No. NBP152179 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152179 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152179 Supplier Novus Biologicals Supplier No. NBP152179
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ATPase Polyclonal specifically detects ATPase in Human, Mouse, Rat, Canine samples. It is validated for Peptide ELISA.

Specifications

Antigen ATPase
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001362.2
Gene Alias ATPase, axonemal, dynein, axonemal, heavy chain 8, dynein, axonemal, heavy polypeptide 8, FLJ25850, FLJ36115, FLJ36334, hdhc9
Gene Symbols DNAH8
Host Species Goat
Immunogen Peptide with sequence C-SQTTHDLSKFDR corresponding to internal region according to NP_001362.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers
Primary or Secondary Primary
Gene ID (Entrez) 1769
Target Species Human, Mouse, Rat, Canine, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.