Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATPase Inhibitory Factor 1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB084392 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB084392 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB084392 Supplier Novus Biologicals Supplier No. NBP299177B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATPase Inhibitory Factor 1 Polyclonal specifically detects ATPase Inhibitory Factor 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ATPase Inhibitory Factor 1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias ATP synthase inhibitor protein, ATPase inhibitor protein, ATPase inhibitor, mitochondrial, ATPase inhibitory factor 1, ATPIIF(1), ATPIP, IF1, Inhibitor of F(1)F(o)-ATPase, IP, MGC1167, MGC8898
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human ATPase Inhibitory Factor 1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 93974
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.