Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATPase Na+/K+ beta 3 Antibody (M016), Novus Biologicals™
SDP

Catalog No. NB387828 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387828 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387828 Supplier Novus Biologicals Supplier No. NBP323433
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATPase Na+/K+ beta 3 Monoclonal antibody specifically detects ATPase Na+/K+ beta 3 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen ATPase Na+/K+ beta 3
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone M016
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence 1:50, Immunoprecipitation 1:50
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias ATPase, Na+/K+ transporting, beta 3 polypeptide, ATPB-3, CD298, CD298 antigen, FLJ29027, Na, K-ATPase beta-3 polypeptide, sodium/potassium-dependent ATPase beta-3 subunit, Sodium/potassium-dependent ATPase subunit beta-3, sodium/potassium-transporting ATPase beta-3 chain, sodium/potassium-transporting ATPase subunit beta-3
Host Species Mouse
Immunogen Clone (M016) was generated from a proprietary antigen related to the native human ATPase Na+/K+ beta 3 subunit expressed in A431 epidermoid carcinoma cell line. [UniProt# P54709]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, CD Markers, Endocrinology, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 483
Target Species Human
Content And Storage Store at -20°C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.