Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATPB Antibody (4.3E8.D10), Novus Biologicals™
SDP

Catalog No. NB6001171 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001171 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001171 Supplier Novus Biologicals Supplier No. NB6001171
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

ATPB Monoclonal specifically detects ATPB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, In vitro assay.

Specifications

Antigen ATPB
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, In vitro Assay
Classification Monoclonal
Clone 4.3E8.D10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunoprecipitation 2 - 5 ug, In vitro assay
Gene Accession No. P06576
Gene Alias ATP synthase subunit beta, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide, ATPMB, ATPSBmitochondrial ATP synthase beta subunit, EC 3.6.3, EC 3.6.3.14, MGC5231, mitochondrial ATP synthetase, beta subunit
Gene Symbols ATP5F1B
Host Species Mouse
Immunogen Intact rat mitochondria.
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 506
Test Specificity Detects the beta subunit of ATP synthase from mouse rat and human samples. This is useful as a mitochondrial marker.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.