Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATPB Antibody (OTI4E5), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB270224APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270224APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270224APC Supplier Novus Biologicals Supplier No. NBP270224APC

Mouse Monoclonal Antibody

ATPB Monoclonal specifically detects ATPB in Human, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen ATPB
Applications Flow Cytometry
Classification Monoclonal
Clone OTI4E5
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ATP synthase subunit beta, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide, ATPMB, ATPSBmitochondrial ATP synthase beta subunit, EC 3.6.3, EC 3.6.3.14, MGC5231, mitochondrial ATP synthetase, beta subunit
Gene Symbols ATP5F1B
Host Species Mouse
Immunogen Full length recombinant protein of human ATPB (NP_001677) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 506.0
Target Species Human, Rat, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.