Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATPB Antibody (OTI4E5), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB279275 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB279275 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB279275 Supplier Novus Biologicals Supplier No. NBP270224MFV610

Mouse Monoclonal Antibody

ATPB Monoclonal antibody specifically detects ATPB in Human, Rat, Canine, Monkey, Primate samples. It is validated for Flow Cytometry

Specifications

Antigen ATPB
Applications Flow Cytometry
Classification Monoclonal
Clone OTI4E5
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias ATP synthase subunit beta, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide, ATPMB, ATPSBmitochondrial ATP synthase beta subunit, EC 3.6.3, EC 3.6.3.14, MGC5231, mitochondrial ATP synthetase, beta subunit
Host Species Mouse
Immunogen Full length human recombinant protein of human ATP5B (NP_001677) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 506
Target Species Human, Rat, Canine, Monkey, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.