Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATPB Rabbit anti-Human, Clone: JM10-90, Novus Biologicals™
SDP

Catalog No. NBP267171 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267171 Supplier Novus Biologicals Supplier No. NBP267171

Rabbit Monoclonal Antibody

ATPB Monoclonal specifically detects ATPB in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen ATPB
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JM10-90
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias ATP synthase subunit beta, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide, ATPMB, ATPSBmitochondrial ATP synthase beta subunit, EC 3.6.3, EC 3.6.3.14, MGC5231, mitochondrial ATP synthetase, beta subunit
Gene Symbols ATP5F1B
Host Species Rabbit
Immunogen Synthetic peptide within Human ATPB aa 141-184/ 529. (SwissProt: P06576 Human; SwissProt: P56480 Mouse; SwissProt: P10719 Rat)
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 506
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.