Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATPG Antibody (OTI2H6), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP270233A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP270233A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP270233A32 Supplier Novus Biologicals Supplier No. NBP270233AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATPG Monoclonal specifically detects ATPG in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ATPG
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2H6
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ATP synthase gamma chain, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1, ATP5CATP synthase subunit gamma, mitochondrial, ATP5CL1mitochondrial ATP synthase, gamma subunit 1, F-ATPase gamma subunit
Gene Symbols ATP5F1C
Host Species Mouse
Immunogen Full length recombinant protein of human ATPG (NP_001001973) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 509.0
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.