Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATR Antibody (2B5), Novus Biologicals™
SDP

Catalog No. NB100308 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100308 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100308 Supplier Novus Biologicals Supplier No. NB100308
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 19 publications

ATR Monoclonal specifically detects ATR in Human, Mouse samples. It is validated for Functional Assay, Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, In vitro Assay, Western Blot.

Specifications

Antigen ATR
Applications Functional Assay, Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, In vitro Assay, Western Blot
Classification Monoclonal
Clone 2B5
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional, In vitro assay, Knockout Validated
Gene Accession No. Q13535
Gene Alias ataxia telangiectasia and Rad3 related, Ataxia telangiectasia and Rad3-related protein, EC 2.7.11.1, FRAP-related protein 1, FRP1FRAP-related protein-1, MEC1, protein kinase ATR, Rad3 related protein, SCKL1, SCKLMEC1, mitosis entry checkpoint 1, homolog, serine/threonine-protein kinase ATR
Gene Symbols ATR
Host Species Mouse
Immunogen Recombinant fragment, corresponding to amino acids 710-1100 of Human ATR.
Molecular Weight of Antigen 317 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Immunology, p53 Pathway, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 545
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.