Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATRAID Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ATRAID |
---|---|
Concentration | 0.17 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4957
![]() ![]() |
Proteintech Group Inc
255481AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody recognizes all the three isoforms (20-24 kDa, 16-19 kDa and 28-30 kDa).
Promotes osteoblast cell differentiation and terminal mineralization. Plays a role in inducing the cell cycle arrest via inhibiting CCND1 expression in all-trans-retinoic acid (ATRA) signal pathway.Specifications
ATRAID | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q6PGD0, Q6UW56 | |
381629, 51374 | |
ATRAID Fusion Protein Ag22168 | |
Primary | |
-20°C | |
ATRAID |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Apoptosis related protein 3, APR 3, APR 3, APR3, C2orf28, HSPC013, p18, PRO240, UNQ214/PRO240 | |
ATRAID | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title