Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atrial Natriuretic Peptide/ANP Antibody (23/1), Novus Biologicals™
SDP

Catalog No. NB036552 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036552 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036552 Supplier Novus Biologicals Supplier No. NBP288961

Mouse Monoclonal Antibody

Atrial Natriuretic Peptide/ANP Monoclonal specifically detects Atrial Natriuretic Peptide/ANP in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Atrial Natriuretic Peptide/ANP
Applications Immunohistochemistry
Classification Monoclonal
Clone 23/1
Conjugate Unconjugated
Formulation PBS
Gene Alias ANF, ANPatriopeptin, ATFB6, atrial natriuretic factor, Atrial natriuretic peptide, CDD-ANF, natriuretic peptide A, natriuretic peptide precursor A, PNDcardionatrin, prepronatriodilatin
Host Species Mouse
Immunogen Recombinant human Atrial Natriuretic Peptide/ANP (aa 1-28).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4878
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.