Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Atrial Natriuretic peptide Rabbit anti-Mouse, Rat, Polyclonal, Chemicon™
Rabbit Polyclonal Antibody
$192.34 - $469.82
Specifications
Antigen | Atrial Natriuretic peptide |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AB5490-I-25UL
|
MilliporeSigma
AB5490I25UL |
25 μL |
Each for $192.34
|
|
|||||
AB5490-I-100
|
MilliporeSigma
AB5490I100UL |
100 μL |
Each for $469.82
|
|
|||||
Description
Anti-Atrial Natriuretic peptide is a highly specific rabbit polyclonal antibody, that targets Natriuretic peptides A and has been tested in Western Blotting and Immunohistochemistry (Paraffin).Specifications
Atrial Natriuretic peptide | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Mouse, Rat | |
Nppa | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Please refer to lot specific datasheet. | |
Polyclonal | |
Rabbit | |
Neuroscience | |
NP_006163.1 | |
KLH-conjugated linear peptide corresponding to 28 amino acids from the C-terminal region of rat Natriuretic peptides A. | |
Affinity Purified | |
This rabbit polyclonal antibody detects Natriuretic peptides A in rat and murine heart. It targets an epitope within 28 amino acids from the C-terminal region. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title