Learn More
Description
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence
Specifications
Specifications
| Antigen | ATRX |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 4212 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 |
| Formulation | PBS with No Preservative |
| Gene Alias | alpha thalassemia/mental retardation syndrome X-linked, alpha thalassemia/mental retardation syndrome X-linked (RAD54 (S. cerevisiae)homolog), alpha thalassemia/mental retardation syndrome X-linked (RAD54 homolog, S.cerevisiae), ATP-dependent helicase ATRX, ATR2, DNA dependent ATPase and helicase, EC 3.6.1, EC 3.6.4.12, helicase 2, X-linked, Juberg-Marsidi syndrome, MGC2094, MRXHF1, RAD54 homolog, RAD54L, SFM1, SHS, transcriptional regulator ATRX, XH2RAD54, X-linked helicase II, X-linked nuclear protein, XNPZNF-HX, Zinc finger helicase, Znf-HX |
| Gene Symbols | ATRX |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
