Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATRX Antibody (rATRX/3446), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB379190 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379190 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379190 Supplier Novus Biologicals Supplier No. NBP308673JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATRX Monoclonal antibody specifically detects ATRX in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen ATRX
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rATRX/3446
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias alpha thalassemia/mental retardation syndrome X-linked, alpha thalassemia/mental retardation syndrome X-linked (RAD54 (S. cerevisiae)homolog), alpha thalassemia/mental retardation syndrome X-linked (RAD54 homolog, S.cerevisiae), ATP-dependent helicase ATRX, ATR2, DNA dependent ATPase and helicase, EC 3.6.1, EC 3.6.4.12, helicase 2, X-linked, Juberg-Marsidi syndrome, MGC2094, MRXHF1, RAD54 homolog, RAD54L, SFM1, SHS, transcriptional regulator ATRX, XH2RAD54, X-linked helicase II, X-linked nuclear protein, XNPZNF-HX, Zinc finger helicase, Znf-HX
Host Species Mouse
Immunogen Recombinant full-length human ATRX protein (Uniprot: P46100 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 546
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.