Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATXN3L Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ATXN3L |
---|---|
Concentration | 0.65 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4009
![]() ![]() |
Proteintech Group Inc
231661AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
ATXN3L, also named as Ataxin-3-like protein, is a 355 amino acid protein, which contains 1 Josephin domain and 2 UIM (ubiquitin-interacting motif) domains. ATXN3L localizes in the nucleus and as a deubiquitinating enzyme that cleaves both ′Lys-48′-linked and ′Lys-63′-linked poly-ubiquitin chains.Specifications
ATXN3L | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9H3M9 | |
92552 | |
ATXN3L Fusion Protein Ag19611 | |
Primary | |
-20°C | |
ATXN3L |
0.65 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ataxin 3 like, ATX3L, ATXN3L, MJDL, Putative ataxin 3 like protein | |
ATXN3L | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title