Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATXN7L1 Antibody (OTI4F6), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP272464A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272464A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272464A94 Supplier Novus Biologicals Supplier No. NBP272464AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATXN7L1 Monoclonal specifically detects ATXN7L1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ATXN7L1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4F6
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ataxin 7-like 1, ataxin 7-like 4, ataxin-7-like protein 1, Ataxin-7-like protein 4, ATXN7L4, FLJ40255, FLJ58242, KIAA1218MGC10760, MGC33190
Gene Symbols ATXN7L1
Host Species Mouse
Immunogen Full length recombinant protein of human ATXN7L1 (NP_689962) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 222255.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.