Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AU1 Epitope Tag Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB209406 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB209406 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB209406 Supplier Novus Biologicals Supplier No. NB600452AF488
Add to Cart
Add to Cart

Goat Polyclonal Antibody

AU1 Epitope Tag Polyclonal antibody specifically detects AU1 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen AU1 Epitope Tag
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias AU1 epitope tag, DTYRYI epitope tag, DTYRYI tag
Host Species Goat
Immunogen Goats were immunized with AU1 (DTYRYI) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags
Primary or Secondary Primary
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.