Learn More
Description
Specifications
Specifications
| Antigen | AU1 Epitope Tag |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 - 1:3000, ELISA 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:400, Immunoprecipitation 1 - 4 ug/mg lysate, Proximity Ligation Assay |
| Formulation | PBS with 0.09% Sodium Azide |
| Gene Alias | AU1 epitope tag, DTYRYI epitope tag, DTYRYI tag |
| Host Species | Goat |
| Immunogen | Goats were immunized with AU1 (DTYRYI) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
