Learn More
Eisco™ Augite (Pyroxene Mineral) Specimen
SCI_ED
Catalog No. p-7076815
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Augite (Pyroxene Mineral) Specimen suitable for use in classroom identification lab exercises and for study on 2-plane, 90° cleavage.
  • For lessons on Bowen's reaction series
  • For mineral identification and physical property exercises
  • Handy visual aid during lessons on Bowen's reaction series as well
  • Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Product Type Mineral, Augite
For Use With (Application) Study on Bowen's reaction series
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.