Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AUH Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB103144 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB103144 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB103144 Supplier Novus Biologicals Supplier No. NBP297719R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AUH Polyclonal specifically detects AUH in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen AUH
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias 3-methylglutaconyl-CoA hydratase, AU RNA binding protein/enoyl-CoA hydratase, AU RNA binding protein/enoyl-Coenzyme A hydratase, AU RNA-binding protein/enoyl-Coenzyme A hydratase, AU-binding protein/enoyl-CoA hydratase, AU-specific RNA-binding enoyl-CoA hydratase, EC 4.2.1.18, methylglutaconyl-CoA hydratase, mitochondrial
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human AUH fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 549
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.