Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AUH Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | AUH |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP157375
![]() |
Novus Biologicals
NBP157375 |
100 μL |
Each for $487.50
|
|
|||||
NBP15737520
![]() |
Novus Biologicals
NBP15737520UL |
20 μL | N/A | N/A | N/A | ||||
Description
AUH Polyclonal specifically detects AUH in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| AUH | |
| Polyclonal | |
| Rabbit | |
| Q13825 | |
| 549 | |
| Synthetic peptides corresponding to AUH (AU RNA binding protein/enoyl-Coenzyme A hydratase) The peptide sequence was selected from the C terminal of AUH. Peptide sequence IGMSLAKELIFSARVLDGKEAKAVGLISHVLEQNQEGDAAYRKALDLARE. | |
| Primary |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| 3-methylglutaconyl-CoA hydratase, AU RNA binding protein/enoyl-CoA hydratase, AU RNA binding protein/enoyl-Coenzyme A hydratase, AU RNA-binding protein/enoyl-Coenzyme A hydratase, AU-binding protein/enoyl-CoA hydratase, AU-specific RNA-binding enoyl-CoA hydratase, EC 4.2.1.18, methylglutaconyl-CoA hydratase, mitochondrial | |
| AUH | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title