Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AUP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501728963 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8963 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8963 Supplier Proteintech Group Inc Supplier No. 137261AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AUP1 contains a domain with homology to the ancient conserved region of the archain 1 gene and a domain thay may be involved in binding ubiquitin-conjugating enzymes. The unprocessed precusor is of 476 amino acids in length and has an estimated molecular weight of 53 kDa.

Specifications

Antigen AUP1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AUP1
Gene Accession No. A1L134, P70295, Q9Y679
Gene Alias ancient ubiquitous protein 1, AUP1
Gene Symbols AUP1
Host Species Rabbit
Immunogen AUP1 Fusion Protein Ag4680
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11993, 550, 680423
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.