Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurora A Antibody (1A14), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB360523 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360523 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360523 Supplier Novus Biologicals Supplier No. NBP250041MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aurora A Monoclonal antibody specifically detects Aurora A in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Aurora A
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1A14
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias AIKAurA, ARK-1, ARK1EC 2.7.11.1, AURA, aurora kinase AhARK1, aurora/IPL1-like kinase, Aurora/IPL1-related kinase 1, Aurora-related kinase 1, Breast tumor-amplified kinase, breast-tumor-amplified kinase, BTAKAURORA2, EC 2.7.11, IPL1-related kinase, serine/threonine kinase 6, serine/threonine protein kinase 15, Serine/threonine-protein kinase 15, serine/threonine-protein kinase 6, Serine/threonine-protein kinase aurora-A, STK15serine/threonine kinase 15, STK6MGC34538, STK7
Host Species Mouse
Immunogen Recombinant full length human Aurora A. [UniProt# O14965]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, MAP Kinase Signaling, Mitotic Regulators, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 6790
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.