Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurora A Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB120222 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120222 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120222 Supplier Novus Biologicals Supplier No. NBP151843JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aurora A Polyclonal specifically detects Aurora A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Aurora A
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias AIKAurA, ARK-1, ARK1EC 2.7.11.1, AURA, aurora kinase AhARK1, aurora/IPL1-like kinase, Aurora/IPL1-related kinase 1, Aurora-related kinase 1, Breast tumor-amplified kinase, breast-tumor-amplified kinase, BTAKAURORA2, EC 2.7.11, IPL1-related kinase, serine/threonine kinase 6, serine/threonine protein kinase 15, Serine/threonine-protein kinase 15, serine/threonine-protein kinase 6, Serine/threonine-protein kinase aurora-A, STK15serine/threonine kinase 15, STK6MGC34538, STK7
Host Species Rabbit
Immunogen A recombinant protein made to an N-terminal region of the human Aurora A protein (within residues 50-200). [Swiss-Prot O14965]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, MAP Kinase Signaling, Mitotic Regulators, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 6790
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.